55324-U
Supel™Genie HybridSPE® Online SPE Starter Kit
for use with Supel™Genie Online SPE Cartridges, includes holder and 1 cartridge, 1 ea of (holder hardware and cartridge)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NB.21
Recommended Products
product line
Supel™
packaging
1 ea of (holder hardware and cartridge)
technique(s)
solid phase extraction (SPE): suitable
L × I.D.
20 mm × 4.0 mm
matrix active group
zirconia-based phase
application(s)
food and beverages
General description
Supel™ Genie HybridSPE® online cartridges have been developed as an alternative to traditional 96-well plate and individual cartridge formats for phospholipid removal and sample preparation. The HybridSPE online cartridges have been designed for online phospholipid removal allowing direct injection of protein precipitated samples for LC/MS analysis. When the samples flow through the HybridSPE online traps, the phospholipids are retained while the analytes of interest flow through.
For first time use, this starter kit is required to obtain reusable universal hardware that will fit all Supel Genie Online SPE cartridges.
LC instrumentation must be configured to accommodate Online SPE cartridges. Please see instrument setup diagram in data sheet 55324-U
Please contact [email protected] for more information on setup and hardware.
For first time use, this starter kit is required to obtain reusable universal hardware that will fit all Supel Genie Online SPE cartridges.
LC instrumentation must be configured to accommodate Online SPE cartridges. Please see instrument setup diagram in data sheet 55324-U
Please contact [email protected] for more information on setup and hardware.
Application
Sample Preparation, HPLC
Features and Benefits
- Provides cleaner samples for longer column life, less instrument downtime, and more accurate and reproducible data
- Automation saves time, decreases costs, and eliminates human error
- Unique Lewis acid/base retention mechanism effectively remove >95% of phospholipids from both hydrophilic and hydrophobic analytes of interest (unlike competitive chemistries) eliminating phospholipid-induced ion suppression
- Highly reproducible, consistent product performance
- Starter kit comes with reusable universal hardware (fits any of the Supel Genie Online SPE cartridges) and 1 cartridge
- Additional cartridges are also available in packs of 2 or 6
Legal Information
HybridSPE is a registered trademark of Merck KGaA, Darmstadt, Germany
Supel is a trademark of Sigma-Aldrich Co. LLC
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Solid phase extraction purifies samples for chromatographic analysis, improving data quality and protecting analytical columns.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service