Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55209

Supelco

Frit Cap Assembly for Supelco® HPLC Column Hardware

size 4.0-4.6 mm, pore size 0.5 μm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

material

PEEK with stainless steel frit

feature

color-coded black

packaging

pkg of 2 ea

size

4.0-4.6 mm

pore size

0.5 μm

compatibility

for use with Supelco 4.0-4.6mm I.D. Discovery and Supelcosil columns
use to protect 3μm packing material

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service