Key Documents
55074
Upchurch Tubing Adapter
About This Item
UNSPSC Code:
41115700
material
PEEK adapter (one-piece design)
packaging
pkg of 1 ea
fitting
male Luer (lock hub-quick connect)
M6 female
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)