Tubing adaptor is a national pipe thread taper made up of stainless steel which is used to connect a female adaptor. It can be used to connect capillary tubes in HPLC, GC.
Legal Information
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Food additives and contaminants, 21(11), 1115-1124 (2005-03-15)
Migration of aniline from polyamide cooking utensils was investigated. Aniline was found to originate from the application of black colorant in the polyamide 66 raw material. Analysis of polyamide raw material revealed an aniline concentration of 121 +/- 13 mg
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.