Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

54989

Kel-F Tee Connector

for use with 1/16 in. tubing (0.25mm bore)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

material

CTFE unit

packaging

pkg of 1 ea

manufacturer/tradename

VICI Valco® CTCKF

bore diam.

0.25 mm

fitting

1/4-28 thread for 1/16 in. O.D. tubing (flangeless fittings included)

compatibility

for use with 1/16" tubing

Looking for similar products? Visit Product Comparison Guide

General description

Connects any three pieces of tubing, using 1/4-28 fittings. Flangeless fittings included.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Valco is a registered trademark of Valco Instruments Co. Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service