Key Documents
54985-U
Tubing Adapter
1/4 in. male NPT, 1/4-28 male Valco
About This Item
UNSPSC Code:
41121815
NACRES:
SB.52
material
CTFE unit
description
for chromatography applications
packaging
pkg of 1 ea
manufacturer/tradename
VICI Valco® CAPA4KF
fitting
1/4-28 male Valco
1/4 in. male NPT
General description
Tubing adaptor is a national pipe thread taper made up of stainless steel which is used to connect a female adaptor. It can be used to connect capillary tubes in HPLC, GC and other chromatographic applications.
Application
Tubing adaptor was used in solid phase extraction.[1]
Legal Information
Valco is a registered trademark of Valco Instruments Co. Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
Customers Also Viewed
C Brede et al.
Food additives and contaminants, 21(11), 1115-1124 (2005-03-15)
Migration of aniline from polyamide cooking utensils was investigated. Aniline was found to originate from the application of black colorant in the polyamide 66 raw material. Analysis of polyamide raw material revealed an aniline concentration of 121 +/- 13 mg