Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

54966

Supelco

LiChrosorb® RP-8 (5 µm) HPLC Columns

L × I.D. 1 cm × 4.6 mm, HPLC Guard Cartridge

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

Product Name

LiChrosorb® RP-8 HPLC Guard Cartridge, 5 μm particle size, L × I.D. 1 cm × 4.6 mm

agency

suitable for USP L7

feature

endcapped: no

packaging

pkg of 4 ea

extent of labeling

10% Carbon loading

technique(s)

HPLC: suitable

L × I.D.

1 cm × 4.6 mm

matrix

Octyl

matrix active group

C8 (octyl) phase

particle size

5 μm

pore size

100 Å

compatibility

use to protect LiChrosorb RP-8

separation technique

reversed phase

General description

Use guard cartridge holder 54987

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChrosorb is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service