Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

54917

Supelco

Nucleosil® C18 HPLC Column

3 μm particle size, L × I.D. 10 cm × 3.2 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

agency

suitable for USP L1

form

solid

feature

endcapped

manufacturer/tradename

Nucleosil®

extent of labeling

15% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

10 cm × 3.2 mm

matrix

octadecyl

matrix active group

C18 (octadecyl) phase

particle size

3 μm

pore size

100 Å

separation technique

reversed phase

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Nucleosil is a registered trademark of Macherey-Nagel GmbH & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service