Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

54904

Supelco

Spherisorb® ODS2 HPLC Column

5 μm particle size, L × I.D. 15 cm × 3 mm

Synonym(s):

WATERS Spherisorb® ODS-2, 5UM, 15CM X 3.2MM ID

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

agency

suitable for USP L1

feature

endcapped

manufacturer/tradename

Spherisorb®

extent of labeling

11.5% Carbon loading

parameter

≤ °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

15 cm × 3 mm

matrix

octadecyl

matrix active group

C18 (octadecyl) phase

particle size

5 μm

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Spherisorb® ODS2 HPLC Columns are packed with the finest commercially available silicas and bonded phases from the most recognized HPLC manufacturers in the world. The columns are tested with specially selected solutes that can be used to establish the reproducibility of retention and selectivity needed in the HPLC applications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Spherisorb is a registered trademark of Waters

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service