Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

54840-U

HiTrap® Protein G Cartridge

part volume 1 mL

Synonym(s):

HiTrap® Affinity Cartridge

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817

packaging

pkg of 5 ea

manufacturer/tradename

Amersham Biosciences HiTrap® 17-0404-01
Amersham Pharmacia Bio

compatibility

mode of use Affinity

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

HiTrap adapters and instruction manual included.

Application

HiTrap® Protein G Cartridges are used in antibody purification and characterization, protein A, G, and L resins and in supplementary products. HiTrap® Protein G columns can be used in affinity chromatography studies isolating monoclonal antibodies.

Legal Information

HiTrap is a registered trademark of Cytiva

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

S B Yan et al.
Journal of thrombosis and haemostasis : JTH, 7(11), 1787-1794 (2009-12-18)
Drotrecogin alpha (activated) (DAA) is a recombinant human activated protein C (APC), which is an antithrombotic protein. To evaluate the development of anti-APC antibodies in severe sepsis patients in DAA clinical studies. Serum and plasma samples were collected in placebo-controlled
Production, characterization and applications of mouse anti-grass carp (Ctenopharyngodon idellus) growth hormone monoclonal antibodies
Yiu-Kwong Leung, M. and W. Kwok-Keung Ho
Comparative Biochemistry and Physiology, 143, 107-115 (2006)
Monoclonal antibodies to plasma high density lipoprotein (HDL) of eel (Anguilla japonica)
Ndiaye, D., et al.
Comparative Biochemistry and Physiology, 125, 473-482 (2000)
C Aybay et al.
Journal of immunological methods, 233(1-2), 77-81 (2000-01-29)
Fetal calf serum (FCS) was depleted of its immunoglobulin G (IgG) in a rapid procedure using protein G affinity chromatography. 20 ml of FCS was depleted of its IgG in less than 80 min by applying 5 ml of FCS

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service