Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

54831

HiTrap® NHS-Activated Cartridge

Synonym(s):

HiTrap® Affinity Cartridge

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
23151817

packaging

pkg of 1 ea

manufacturer/tradename

Amersham Biosciences HiTrap® 17-0717-01
Amersham Pharmacia Bio

compatibility

mode of use Affinity

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

HiTrap adapters and instruction manual included.

Application

HiTrap® NHS-Activated Cartridges are used in affinity chromatography, protein chromatography and activated or functionalized matrices. HiTrap® NHS-Activated columns have been used in studies assessing the generation and quantitation of antibodies, analysis of protein expression and peptide synthesis.

Legal Information

HiTrap is a registered trademark of Cytiva

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

53.6 °F - closed cup

flash_point_c

12.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peptide synthesis on Sepharose beads
Tegge, W. & R. Frank
The Journal of Peptide Research, 49, 255-262 (1997)
Qing-Guo Zhao et al.
Yi chuan xue bao = Acta genetica Sinica, 33(1), 49-55 (2006-02-03)
Fanconi anemia complementation group L (FANCL) is a novel Fanconi anemia protein, which mono-ubiquitinates FANCD2 as a ubiquitin E3 ligase, and plays a crucial role in DNA damage repair and chromosome stability maintenance. FANCL is involved in the proliferation of
N P Koper et al.
Clinical chemistry and laboratory medicine, 36(1), 23-28 (1998-05-22)
Currently no available immunoassay system offers complete protection against spuriously elevated or lowered results due to interference by Human Anti-Mouse Antibodies (HAMA). Although routine use of chromatography procedures is not an acceptable option because of the extra cost and workload

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service