Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

54796-U

Supelco

LiChrospher® NH2 HPLC Guard Cartridge

5 μm particle size, L × I.D. 1 cm × 4.6 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

material

stainless steel column

agency

suitable for USP L8

product line

LiChrospher®

feature

endcapped: no

packaging

pkg of 4 ea

extent of labeling

4.6% Carbon loading

parameter

≤70 °C temp. range

technique(s)

HPLC: suitable

L × I.D.

1 cm × 4.6 mm

surface area

350 m2/g

matrix

Amino
fully porous particle

matrix active group

amino phase

particle size

5 μm

pore size

100 Å

operating pH

2-7.5

application(s)

food and beverages

compatibility

for use with 3.0 to 4.6 mm column

separation technique

anion exchange
hydrophilic interaction (HILIC)
normal phase
reversed phase

General description

Use guard cartridge holder 54987

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChrospher is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service