Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

54784

Supelco

LiChrospher® NH2 (5 µm) HPLC Columns

L × I.D. 25 cm × 4.6 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

Product Name

LiChrospher® NH2 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm

agency

suitable for USP L8

feature

endcapped: no

manufacturer/tradename

LiChrospher®

extent of labeling

4.6% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

surface area

350 m2/g

matrix

amino

matrix active group

amino phase

particle size

5 μm

pore size

100 Å

application(s)

food and beverages

separation technique

anion exchange
hydrophilic interaction (HILIC)
normal phase

Looking for similar products? Visit Product Comparison Guide

General description

LiChrospher® is a reliable and versatile spherical silica sorbent, which is traditionally produced and available with different modifications. LiChrospher® NH2 (5 μm particle size, L × I.D. 25 cm × 4.6 mm) is a highly versatile sorbent with both polar and hydrophobic properties and is suitable for chromatographic separation of carbohydrates (mono-, di- and oligosaccharides) with reversed-phase chromatography, or the separation of nucleotides by ion exchange chromatography.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChrospher is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service