ProtEx columns are 5.0 cm x 4.6 mm I.D. polyetheretherketone (PEEK) hardware packed with high performance, monodisperse, 5 μm, 1000 Å macroporous polymethacrylate beads with a chemically bonded, crosslinked hydrophilic surface.
Features and Benefits
Excellent protein separations
High resolution at low sample load
Quantitative recovery
High efficiency
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
MCI GEL is a registered trademark of Mitsubishi Chemical Corp.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.