Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

54740-U

MCI GEL® ProtEx-SP HPLC Column - Deleted, Substitute p/n 59601-U

5 μm particle size, L × I.D. 5.0 cm × 4.6 mm

Synonym(s):

MCI GEL® Ion Exchange Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

material

PEEK column

manufacturer/tradename

ProtEx-SP

L × I.D.

5.0 cm × 4.6 mm

matrix

polymeric

matrix active group

sulfopropyl

particle size

5 μm

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

ProtEx columns are 5.0 cm x 4.6 mm I.D. polyetheretherketone (PEEK) hardware packed with high performance, monodisperse, 5 μm, 1000 Å macroporous polymethacrylate beads with a chemically bonded, crosslinked hydrophilic surface.

Features and Benefits

  • Excellent protein separations
  • High resolution at low sample load
  • Quantitative recovery
  • High efficiency
   

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

MCI GEL is a registered trademark of Mitsubishi Chemical Corp.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service