Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

54718-U

Upchruch Universal Prime/Purge Valve

low pressure

Synonym(s):

Upchurch Prime/Purge Valve

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
SB.51

material

PEEK/PTFE unit

description

Includes 2 Tefzel®
Ferrules and 2 PEEK nuts for 1/8″ Tubing

Universal Prime/Purge Valve
universal design

fitting

1/4-28 valve ports

Looking for similar products? Visit Product Comparison Guide

General description

Automatic valve operation with a simple twist of the Luer-Lock syringe. Easy installation, long seal life.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Tefzel is a registered trademark of E. I. du Pont de Nemours and Company

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service