Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

54263-U

Supelco

Supel-Tips Ti Pipette Tips

volume 10 μL, pkg of 96 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
eCl@ss:
32079201
volume:
10 μL
packaging:
pkg of 96 ea

material

polypropylene tip

description

Titania bonded on silica, adsorbent

packaging

pkg of 96 ea

volume

10 μL

matrix active group

titania bonded on silica bonding

particle size

50-60 μm

pore size

300 Å pore size

application(s)

food and beverages

Looking for similar products? Visit Product Comparison Guide

General description

Supel-Tips are designed for the micro-scale extraction, purification, and enrichment of femtomole to picomole quantities of biological macromolecules for subsequent analysis and identification with mass spectrometric and/or chromatgraphic techniques.

Supel-Tips contain a chromatographic sorbent bed bonded at the working end of the tip with a proprietary high purity adhesive. The sorbent bed acts as a solid phase extraction medium that adsorbs molecules of interest from the sample matrix. Subsequently, the concentrated and desalted analytes are eluted for downstream analysis.

Application

Supel-Tips Ti Pipette Tips are designed for the extraction of phosphopeptides and other phosphate containing molecules

Features and Benefits

  • Superior recovery
  • Exceptional binding capacity and enhanced affinity
  • Excellent sorbent bed stability for cleaner samples
  • Fast and effective analyte retention/elution

Other Notes

Browse all pipette tips

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service