Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

54254-U

Supelco

Supelpak-4

bed wt. 400 mg, size 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12190000
NACRES:
NB.24

composition

bed wt., 400 mg

packaging

pack of 10

technique(s)

active air sampling: suitable

size

1 mL

application(s)

air monitoring
industrial hygiene

storage temp.

2-8°C

Legal Information

Supelpak is a trademark of Sigma-Aldrich Co. LLC

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F J Egea González et al.
Journal of AOAC International, 80(5), 1091-1097 (1997-11-05)
A method to sample and analyze chlorothalonil and dichlofluanid in greenhouse air was evaluated. Analysis was performed by gas chromatography with electron capture detection and gas chromatography-mass spectrometry. Solid sorbents such as Chromosorb 102, Porapak R, Supelpak-2, Amberlite XAD-2, Amberlite
D Rochat et al.
Journal of chemical ecology, 17(11), 2127-2141 (1991-11-01)
Male American palm weevils (APWs),Rhynchophorus palmarum (L.) produced two sex-specific compounds, which were disclosed by volatile collections on Supelpak-2 and gas chromatography. One was a minor compound, not always detected. The major male-produced volatile was identified as (2E)-6-methyl-2-hepten-4-ol through coupled
Nikolaos G Tsiropoulos et al.
Analytica chimica acta, 573-574, 209-215 (2007-08-29)
A methodology is described for greenhouse air analysis by sampling fenhexamid, pyrimethanil, malathion, metalaxyl-M and myclobutanil in solid sorbents. Pesticides were determined by gas chromatography with NP Detector. The trapping efficiency of XAD-2, XAD-4, Supelpak-2, Florisil and C-18 at different
F J Egea González et al.
Journal of chromatography. A, 829(1-2), 251-258 (1999-01-29)
The diminution of metamidophos residue levels with time in vegetables an greenhouse air was investigated after treatment of tomatoes and green beans. A gas chromatographic method using dichloromethane as an extraction solvent has been developed to analyse metamidophos in vegetables
Olivier Briand et al.
The Science of the total environment, 288(3), 199-213 (2002-05-07)
An analytical method using gas chromatography-mass spectrometry has been developed for the evaluation of different sampling techniques to characterise spray drift in a commercial apple orchard. Eleven pesticides were studied (fungicides, insecticides and herbicides). A collection of airborne spray-drift pesticides

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service