54231-U
Supel™-Select SAX SPE Tube
bed wt. 30 mg, volume 1 mL, pk of 100
About This Item
Recommended Products
material
PE frit (20 μm)
polypropylene hardware
quality
MS Friendly
composition
bed wt., 30 mg
packaging
pk of 100
technique(s)
solid phase extraction (SPE): suitable
surface area
160-420 m2/g
volume
1 mL
matrix
Quaternary amine functionalized hydrophilic modified styrene polymer particle platform
matrix active group
SAX phase
particle size
50-70 μm
pore size
80-200 Å
application(s)
food and beverages
separation technique
ion exchange
Looking for similar products? Visit Product Comparison Guide
General description
Sample Matrix Compatibility: Aqueous solutions (biological fluids, water)
- Quaternary amine functionalized hydrophilic modified styrene polymer
- Developed for the extraction of a highly broad range of compounds from aqueous samples
- Retention predominately based on ion-exchange; however, because the phase is hydrophilic modified, it is amenable to polar compounds as well.
Features and Benefits
- Excellent sample prep performance at a lower price
- Amenable to generic methodology - save time, money, and headache during method development
- Greater capacity allows for smaller bed weights = smaller elution volumes = time savings in sample processing
- Resistance to overdrying allows for more robust methdology
- Low UV and MS extractables for lower background and greater sensitivity
- Stringent production and QC guidelines offer greater lot-to-lot, tube-to-tube, and well-to-well consistency for improved accuracy and precision
Legal Information
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service