Key Documents
53629-U
Supelco
Supel-Connect Agilent® 1100/1200 Interconnect
L × O.D. × I.D. 15 cm × 1/16 in. × 0.005 in.
About This Item
UNSPSC Code:
41115700
type
(rigid)
packaging
pkg of 1 ea
L × O.D. × I.D.
15 cm × 1/16 in. × 0.005 in.
compatibility
HPLC column to heater outlet
General description
For Agilent 1100/1200
The high performance fitting is now available for the Agilent 1100/1200 HPLC system. Semi-rigid 316 stainless steel tubing connects one end to the heater outlet, and allows use of columns by any manufacturer with confidence that the high performance UHPLC fitting is completely seated in the column inlet port. An improtant feature of this fitting is the ′service loop′, which allows the column to be semi-rigdily supported.
Legal Information
Agilent is a registered trademark of Agilent Technologies, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)