53202-U
SupelMIP® SPE - Beta-agonists
bed wt. 25 mg, volume 10 mL , LRC, pk of 50
About This Item
Recommended Products
material
polypropylene tube
product line
SupelMIP®
composition
bed wt., 25 mg
packaging
pk of 50
technique(s)
solid phase extraction (SPE): suitable
volume
10 mL , LRC
matrix active group
SupelMIP® phase
Looking for similar products? Visit Product Comparison Guide
General description
The SupelMIP SPE line consist s of highly cross-linked polymers that are engineered to extract a single analyte of interest or a class of structurally related analytes with an extremely high degree of selectivity. This is possible because selectivity is introduced during MIP synthesis in which a template molecule, designed to mimic the analyte, guides the formation of specific cavities or imprints that are sterically and chemically complementary to the analyte(s) of interest.
By careful design of the imprinting site, either by molecular modeling, experimental design, or screening methods, the binding cavities can be engineered to offer multiple interaction points with the analyte(s) of interest. This leads to a stronger interaction between the solid-phase and the analyte(s). As a consequence, harsher wash conditions can be tolerated during SPE methodology resulting in cleaner extracts. Because extraction selectivity is significantly improved, lower background is observed allowing analysts to achieve lower limits of detection.
Visit www.sigma-aldrich.com/supelmip to learn more.
Application
SupelMIP - Beta-agonists was developed for the selective extaction of a broad range o beta-agonists such as brombuterol, clenbuterol, formoterol, isoxuprine, mapenterol, ractopamine, ritrodine, salbutamol, salmeterol, terbutaline, tulobuterol, zilpaterol and others. The SupelMIP cartridge offers excellent reproducibility for surveillance of this common class of drugs of abuse. Loading capacity is superior to immuno-affinity columns, and MS-detection is recommended for forensic validity. Detailed extraction protocols are included with the product.
Features and Benefits
- Highly selective SPE for trace analysis in complex matrices
- Achieve lower detection limits
- Significant time and cost savings
- Improved MS-compatibility
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
200.3 °F
flash_point_c
93.5 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
SupelMIP SPE is based on molecularly imprinted polymer (MIP) technology. Each SupelMIP phase offers tailor-made selectivity for the extraction of trace analytes in complex matrixes.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service