52576-U
Supelclean™ SAX/PSA SPE Tube
bed A 250 mg (Supelclean™ LC-SAX SPE), bed B 250 mg (Supelclean™ PSA SPE), volume 6 mL, pk of 30
About This Item
Recommended Products
material
PE frit
polypropylene hardware
composition
bed A, 250 mg (Supelclean™ LC-SAX SPE)
bed B, 250 mg (Supelclean™ PSA SPE)
packaging
pk of 30
technique(s)
solid phase extraction (SPE): suitable
volume
6 mL
matrix active group
PSA phase
SAX phase
application(s)
food and beverages
separation technique
ion exchange
normal phase
Looking for similar products? Visit Product Comparison Guide
General description
Sample Matrix Compatibility: Organic or aqueous solutions
- Dual layer SPE tube that contains both Supelclean SAX (upper layer) and PSA (lower layer) SPE sorbents (separated by PE frit)
- Supelclean SAX is a quarternary amine, Cl- counter-ion.
- Supelclean PSA an ethylenediamine-N-propyl phas that contains both primary and secondary amines.
- Ideal for removing matrix components (fatty acids, organic acids, polar pigments, and some sugars) when conducting multi-residue pesticide analysis in foods
- In compliance with the Luke II method which uses SPE to remove matrix interference and enhancement of pesticides from food for GC-ITMS analysis
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Inhalation - STOT SE 3
target_organs
Respiratory system, respiratory tract irritation
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service