Key Documents
52257-U
Biocompatible UHPLC PEEK Lined Stainless Steel Tubing with Fittings
L × O.D. × I.D. 300 mm × 1/16 in. × 100 μm
About This Item
UNSPSC Code:
41115700
packaging
1 ea of
L × O.D. × I.D.
300 mm × 1/16 in. × 100 μm
General description
Upchurch Scientific′s new Biocompatible PEEK-Lined Stainless Steel (PLS) Tubing for biocompatible UHPLC applications. The tubing combines the strength of industry-standard 316 Stainless Steel with the chemical inertness of PEEK polymer to enable more efficient bio-separations at pressures up to 15,000 psi (1,034 bar).
Designed to Work with VHP Fittings
PLS Tubing achieves its maximum performance of 15,000 psi (1,034 bar) when used with Upchurch Scientific VHP Fittings. The standard configuration of this tubing automatically pairs a length of tubing with two VHP-325 fittings, which allow repeat connections at UHPLC pressures.
Strong Yet Flexible
The unique design features of PLS Tubing allow it to be bent into shapes that may be required by the system equipment - including angled bends and even sample loops for the injection valve - all with no loss of performance. Even in a bent shape, the PEEK lining maintains its integrity along the entire length.
Choose from one of the most recent versions:
Certificates of Analysis (COA)