Key Documents
52246-U
Upchurch VHP (Very High Pressure) Inline Filter
VHP-505, porosity 0.2 μm, for use with 1/16 inch tubing, stainless steel
About This Item
UNSPSC Code:
41115700
NACRES:
SB.52
material
stainless steel
manufacturer/tradename
VHP-505
packaging
1 ea of
pore size
0.2 μm porosity
compatibility
for use with 1/16 inch tubing
General description
Inline Between the Pump and Valve
When placed between a high pressure pump and the injection valve in a UHPLC system, the VHP Inline Filter prolongs the life of the valve′s rotor seal by catching particles sloughed off due to pump seal wear. The small volume of the VHP liline Filter helps ensure mobile pbase gradient changes reach the column with minimal delay.
Footnote
This product ships complete with a total of five (5) replacement frits (including the one pre-assembled into the product)
Choose from one of the most recent versions:
Certificates of Analysis (COA)