Key Documents
52223-U
Upchurch VHP (Very High Pressure) Fittings
PK-126, for use with 1/32 inch (0.8mm) tubing, 6-32 thread, (polymer)
About This Item
UNSPSC Code:
41115700
NACRES:
SB.52
material
(polymer)
manufacturer/tradename
PK-126
packaging
1 ea of
fitting
6-32 thread
compatibility
for use with 1/32 inch (0.8mm) tubing
Choose from one of the most recent versions:
Certificates of Analysis (COA)