Key Documents
51901-U
MarvelX UHPLC Connection Systems Replacement Tubing
Stainless Steel, I.D. × L 125 μm × 350 mm
About This Item
UNSPSC Code:
12352200
NACRES:
SB.52
I.D. × L
125 μm × 350 mm
General description
MarvelX UHPLC Connection Systems have been expertly designed for easier routing throughout your instrument, with consistent performance and superior re-usability. Built with convenient, removable stainless steel fitting and changeable precision-cut flexible tubing, MarvelX can be used up to 200 times! This amazing design only requires 2 in-lbs of installation torque, and seals up to 19,000 psi (1,310 bar). MarvelX is compatible with 10-32 coned receiving ports and is truly finger-tight - no tool required.
Legal Information
MarvelX is a trademark of IDEX Health & Science, LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)