Key Documents
51367-U
Supelco
Supel-Connect High Performance Fittings
Long Fitting with Ferrules, pack of 2
About This Item
UNSPSC Code:
41115700
packaging
pack of 2
compatibility
for use with PEEK/PEEKSil tubing
Features and Benefits
  • Fitting are for use with PEEK/PEEKSil or stainless steel tubing
  • Used at pressures to 15,000 psi
  • SupelConnect Better Nut allows use in tighter places
  • SupelConnect Long and Short High Performance fitting adjust to any port depth
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Supel is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)