Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51328-U

Supelco

PEEKsil Tubing

black, pkg of 2 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

packaging

pkg of 2 ea
pkg of

color

black

Looking for similar products? Visit Product Comparison Guide

General description

PEEKsil is fused silica tubing sheathed in PEEK polymer, with an effective outer diameter. The PEEK sheating is mechanically strong and has ideal characteristics for sealing with metal or polymer fittings. The fused silica core provides a consistent and rigid fluid pathway with very tight tolerances and industry-accepted chemical properties. Like traditional fused silica tubing, PEEKsil has excellent chemical compatibility and extremely low absorption characteristics, especiall when compared to stainless steel.

Please note: Do not cut this tubing. It should be used as its precut lengths because of permanent damage caused by conventional cutters.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

PEEKSil is a trademark of SGE Analytical Science Pty Ltd.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service