Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51274-U

Ultra-High Performance Micro Fingertight Unions

for use with 1/32" Tubing, pkg of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

description

PEEK fittings and stainless steel body
includes 2 MicroFerrules for 1/32" O.D. tubing and 2 female 5/16-24 internal thread PEEK nuts

packaging

pkg of 1 ea

compatibility

for use with 1/32" Tubing

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service