Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51256-U

Ultra-High Performance Fingertight Fittings

PEEK (LiteTouch® Nut (Black), 10-32), for use with 1/32" tubing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

material

PEEK (LiteTouch® Nut (Black), 10-32)

description

Note: ferrules sold separaely (requires 51257-U)

packaging

pkg of 10 ea

compatibility

for use with 1/32" tubing

Looking for similar products? Visit Product Comparison Guide

General description

The innovative line of Upchurch Ultra High Performance fittings are desigined to withstand extreme temperature anf pressures. This line of fittings is perfect for use within the increasingly demanding requirements for today′s high performance analytical systems. The fittings are a 10/32 fingertight style for use with 1/16" and 1/32" OD tubing.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiteTouch is a registered trademark of IDEX Health & Science, LLC

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service