Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51245-U

Nonmetallic 1/4-28 and Inline Cartridge Check Valves

inlet check valve,1/4"-28 male to 1/4"- 28 female, pkg of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

packaging

pkg of 1 ea

Looking for similar products? Visit Product Comparison Guide

General description

These check valves function well in both low and high pressure applications. Low internal volume also allows them to be used in areas where flow path volume is important; however, higher flows can pass through with minimal pressure drop. Metal-free composition makes the check valve perfect for use with corrosive fluids or biological samples.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service