Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

509981

Supelco

EPA TO-14 Calibration Mix 1 (39 components)

1 ppm in nitrogen, pkg of 110 L (pi-marked cylinder), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41121813

grade

analytical standard

agency

EPA TO-14,TO-15,TO-17

packaging

pkg of 110 L (pi-marked cylinder)

manufacturer/tradename

Scott Gas T9Z90001-110

concentration

1 ppm in nitrogen

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

Looking for similar products? Visit Product Comparison Guide

General description

Air Liquie America Specialty Gases LLC TO-14 gas blends are packaged in SCOTTY high pressure, transportable, laboratory-size cylinders. A SCOTTY cylinder contains over 100 liters of gas at 1800psi--enough gas to deliver adequate calibration for 80 or more uses. Each cylinder is equipped with CGA 180 fittings to plumb directly to your gas sampling system or through an appropriate regulator.

Where available, the components in Scott TO-14 gas calibration blends are traceable to NIST reference mixtures (NIST SRM 1804). All are certified for superior stability and accuracy.

The shelf life is one year.

Application

EPA TO-14 calibration mix 1 (39 components) may be used as a gas standard for the calibration and validation of a portable gas chromatography coupled with mass spectrometry system (GC-MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Benzene
    Bromomethane
    Carbon tetrachloride
    Chlorobenzene
    Chloroethane
    Chloroform
    Chloromethane
    1,2-Dibromoethane
    1,2-Dichlorobenzene
    1,3-Dichlorobenzene
    1,4-Dichlorobenzene
    Dichlorodifluoromethane
    1,1-Dichloroethane
    1,2-Dichloroethane
    1,1-Dichloroethylene
    cis-1,2-Dichloroethylene
    Dichloromethane
    1,2-Dichloropropane
    cis-1,3-Dichloropropene
    trans-1,3-Dichloropropene
    1,2-Dichlorotetrafluoroethane
    Ethylbenzene
    Hexachloro-1,3-butadiene
    Styrene
    1,1,2,2-Tetrachloroethane
    Tetrachloroethylene
    Toluene
    1,2,4-Trichlorobenzene
    1,1,1-Trichloroethane
    1,1,2-Trichloroethane
    Trichloroethylene
    Trichlorofluoromethane
    1,1,2-Trichloro-1,2,2-trifluoroethane
    1,2,4-Trimethylbenzene
    1,3,5-Trimethylbenzene
    Vinyl chloride
    o-Xylene
    m-Xylene
    p-Xylene
    See All (39)

regulator

Product No.
Description
Pricing

pictograms

Gas cylinder

signalword

Warning

hcodes

Hazard Classifications

Press. Gas Compr. Gas

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Equilibrium distribution sampling device for preparation of calibration mixtures for gas chromatography-mass spectrometry
Xie X, et al.
Analytical Methods : Advancing Methods and Applications, 22(5), 6312-6318 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service