Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

509876

SGE Microvolume Syringe for Rheodyne® and Valco® Valves, Removable Needle

25R-GT-LC, volume 25 μL, needle size 22 ga, needle L × O.D. 2 in. × 0.7112 mm (0.028 in.)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001

material

blunt tip stainless steel needle

needle size

22 ga

feature

needle type RN
point style #3 (blunt tip)
syringe type LC
Removable needle

packaging

pkg of 1 ea

manufacturer/tradename

SGE 003312

needle L × O.D.

2 in. × 0.7112 mm (0.028 in.)

volume

25 μL

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Rheodyne is a registered trademark of IDEX Health & Science, LLC
Valco is a registered trademark of Valco Instruments Co. Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service