Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

50941-U

Zorbax® HPLC Column

phase cyano, 5 μm particle size, L × I.D. 25 cm × 4.6 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

product line

Zorbax®

feature

endcapped: no

manufacturer/tradename

Zorbax®

extent of labeling

7% loading

parameter

≤70 °C temp. range

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

surface area

330 m2/g

matrix

cyano

matrix active group

cyano phase

particle size

5 μm

pore size

70 Å

application(s)

food and beverages

separation technique

normal phase

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Zorbax is a registered trademark of Agilent Technologies, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service