Key Documents
508187
Supelco
Vial crimp Seal with Red Rubber/PTFE Septum
seal diam. 11 mm, blue aluminum, PTFE/red rubber, pkg of 100 ea
About This Item
UNSPSC Code:
41122001
material
PTFE/red rubber
blue aluminum
packaging
pkg of 100 ea
seal diam.
11 mm
Choose from one of the most recent versions:
Certificates of Analysis (COA)