Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

507113

Supelco

Vacuum Sampling Pumps

Model 1067 Tube Sampler (Dual channel)

Synonym(s):

Air Sampling Pumps

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008

technique(s)

whole air sampling: suitable

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with Sorbent Tubes (ORBO and TD)

Looking for similar products? Visit Product Comparison Guide

Other Notes

A universal battery charger (110V/240V) is included with part number 507113. This universal charger (24697-U) can be ordered separately, if a replacement is required.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service