Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

50684-U

Hamilton PRP-C18 Analytical Guard Column Starter Kit

5 μm particle size, L × I.D. 20 mm × 2 mm, PEEK, kit of

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

material

PEEK

manufacturer/tradename

Hamilton

packaging

kit of

technique(s)

HPLC: suitable

L × I.D.

20 mm × 2 mm

matrix

PS-DVB particle platform
polymeric

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

100 Å

operating pH

1-14

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Kit includes 1 holder and 2 cartridges

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service