Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

506818

Supelco

Ethanethiol (ethyl mercaptan)

PESTANAL®, analytical standard

Synonym(s):

Ethanethiol, Ethyl mercaptan, Mercaptan C2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5SH
CAS Number:
Molecular Weight:
62.13
Beilstein/REAXYS Number:
773638
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

2.1 (vs air)

vapor pressure

8.51 psi ( 20 °C)

product line

PESTANAL®

CofA

current certificate can be downloaded

autoignition temp.

570 °F

expl. lim.

18.2 %

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.4306 (lit.)

bp

35 °C (lit.)

density

0.839 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCS

InChI

1S/C2H6S/c1-2-3/h3H,2H2,1H3

InChI key

DNJIEGIFACGWOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-49.0 °F - closed cup

flash_point_c

-45 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Hanczkó et al.
Journal of chromatography. A, 1163(1-2), 25-42 (2007-07-04)
The main aims of this work were (a) to present the characteristics and stability of the o-phthalaldehyde (OPA)-ethanethiol (ET) derivatives of 22 amino acids, including the believed-to-be less stable OPA derivatives providing glycine, gamma-aminobutyric acid, beta-alanine, histidine, ornithine, lysine and
Shungang Wan et al.
Biodegradation, 21(6), 1057-1066 (2010-05-15)
In the present study, a new bacterial strain isolated from activated sludge has been identified as Lysinibacillus sphaericus based on its morphology, physiochemical properties, and the results of 16S ribosomal RNA (rRNA) gene sequence analysis. This new bacterial strain uses
J A Gaskin et al.
Rhinology, 46(3), 166-169 (2008-10-16)
To determine if 1) there is cross contamination between odours tested on thresholds achieved, 2) a delay period is necessary between testing different odours. Thirty-five subjects underwent threshold testing with phenethyl alcohol (PEA), ethylmercaptan (MER), acetic acid (ACE), and eucalyptol
Taicheng An et al.
Journal of hazardous materials, 183(1-3), 372-380 (2010-08-10)
This study aims to compare the biological degradation performance of ethanethiol using strain RG-1 and B350 commercial mixed microorganisms, which were inoculated and immobilized on ceramic particles in twin-biotrickling filter columns. The parameters affecting the removal efficiency, such as empty
Jiangyao Chen et al.
Journal of hazardous materials, 190(1-3), 416-423 (2011-04-20)
A series of adsorptive photocatalysts, combined titania-montmorillonite-silica were synthesized. The resultant photocatalysts consisted of more and more spherically agglomerated TiO(2) particles with increasing of TiO(2) content, and anatase was the only crystalline phase with nano-scale TiO(2) particles. With increasing of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service