Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

506753

Supelco

ASTM® D5134 Splitter Linearity Check Mix

10 % (w/w) each component (no solvent), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

analytical standard

agency

ASTM® D5134

CofA

current certificate can be downloaded

packaging

pkg of 1 mL

concentration

10 % (w/w) each component (no solvent)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-30°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Neat at 10% by weight, each component. Use this quantitative mix of hydrocarbons to determine proper system performance and detector response.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

    Benzene
    2,4-Dimethylheptane
    2,4-Dimethylhexane
    Heptane
    Hexane
    2-Methylheptane
    2-Methylhexane
    Nonane
    Octane
    Toluene
    See All (10)

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT RE 2 - STOT RE 2 Inhalation - STOT SE 3

target_organs

Blood, Central nervous system, Nervous system, Respiratory system

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service