Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

505846

Supelco

Silylation Sampler Kit

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200

storage temp.

2-8°C

Kit Components Only

Product No.
Description

  • BSA 3 x 1

  • BSTFA 3 x 1

  • BSTFA + TMCS, 99:1 (Sylon BFT) 3 x 1

  • HMDS + TMCS, 3:1 (Sylon HT) 3 x 1

  • TMSI 3 x 1

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

111.2 °F - closed cup

flash_point_c

44 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service