Key Documents
505765
Shutoff Valve, Low Pressure
blue Tefzel®, for use with 1/16 in. tubing
About This Item
UNSPSC Code:
41115854
material
blue Tefzel®
description
manual shut-off valve
feature
positions 2 (on and off)
manufacturer/tradename
Upchurch Scientific, Inc P-782
parameter
600 psi max. pressure
fitting
1/4-28 female in-line for 1/16 in. O.D. tubing
compatibility
for use with 1/16 in. tubing
General description
  • Designed for lower pressure applications
  • Biocompatible, all polymer flow path
PEEK or Tefzel valve with 1/4-28 fittings, for pressures up to 1000 psi (70 kg/cm2). Flangeless nuts and ferrules included.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)