Key Documents
505757
Sigma-Aldrich
Shutoff Valve, Low Pressure
natural PEEK, for use with 1/16 in. tubing
About This Item
UNSPSC Code:
41113000
eCl@ss:
32060101
material
natural PEEK
description
manual shut-off valve
feature
positions 2 (on and off)
manufacturer/tradename
Upchurch Scientific, Inc P-732
parameter
500 psi max. pressure
fitting
1/4-28 female Upchurch in-line for 1/16 in. O.D. tubing
compatibility
for use with 1/16 in. tubing
General description
  • Designed for lower pressure applications
  • Biocompatible, all polymer flow path
PEEK or Tefzel valve with 1/4-28 fittings, for pressures up to 1000 psi (70 kg/cm2). Flangeless nuts and ferrules included.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)