description
two pigtails (on brackets with CGA connectors)
feature
Two station
fitting
CGA 590
General description
- CGA 580 for helium, nitrogen, and argon
- CGA 350 for hydrogen, ethane, and 5% methane in argon
- CGA 590 for compressed air
The two station bracket has 2 pigtails that join before the regulator fitting, and allows 2 gas cylinders to feed the system, extending the time between cylinder change-out.
Choose from one of the most recent versions:
Certificates of Analysis (COA)