503576
Automatic Changeover Panel
CGA 350, for use with H2, Ar/CH4
About This Item
Recommended Products
fitting
CGA 350
compatibility
for use with H2, Ar/CH4
General description
Note: This unit contains a 1-stage regulator, and requires that an in-line pressure regulator is installed downstream for final pressure control.
Specifications:
- Check valve in inlets; keeps air from entering during cylinder change-out
- Stainless steel diaphragm; appropriate for most applications
- Diaphragm packless purge valves; higher leak integrity than needle valve
- Shut-off valve at unit outlet; keeps air from entering during change-out of downstream components
Choose from one of the most recent versions:
Certificates of Analysis (COA)