Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

502936

Supelco

Mounting Clip

for use with 100 cc traps, pkg of 1 ea

Synonym(s):

Clip for 100cc CO2 Trap

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121813

packaging

pkg of 1 ea

compatibility

for use with 100 cc traps

Looking for similar products? Visit Product Comparison Guide

General description

This clip is designed to fit one of our in-line traps based on the volume of the trap. Simply mount two or more clips to a bench or wall using screws (not supplied), then snap the trap into the clips. One plastic clip per package.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service