Key Documents
502804
Upchurch Microtight® Fittings Kit
About This Item
UNSPSC Code:
41115700
General description
Kit includes 2 Microtight unions, 2 Microtight adapters, and various Microtight tubing sleeves. The kit includes 10 each of 8 sizes of Microtight tubing sleeves, ranging in I.D. from 125 μm - 530 μm (color-coded). The appropriate sized tubing sleeve has an I.D. 0.002 in. to 0.003 in. greater than the O.D. of your tubing.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Microtight is a registered trademark of IDEX Health & Science, LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)