Key Documents
502480
MicroCross
for use with MicroTight sleeves, pkg of 1 ea
About This Item
UNSPSC Code:
41121815
description
Connects 4 pieces of 0.025″ O.D. capillary tubing
includes 4, microferrules for .025″ O.D. tubing and 4, female nuts (5/16"-24 internal threads)
packaging
pkg of 1 ea
thru-hole diam.
0.006 in. (150 μm)
compatibility
for use with MicroTight sleeves
General description
Connects 4 capillary tubes, using MicroFingertight II fittings. Includes 4 MicroFerrules for 0.025 in. O.D. tubing, 4 female nuts.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)