Key Documents
502421
Upchurch PEEK Microtight® Connector
Connects 1/16″ capillary tubing
Synonym(s):
Microtight® Adapter
About This Item
UNSPSC Code:
41115700
General description
Connects capillary tubing to 1/16 in. O.D. tubing. True zero-dead volume design. Use gauge plug included) to properly seat tubing. Includes 1 MicroFingertight fitting, 1 standard Fingertight fitting.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Microtight is a registered trademark of IDEX Health & Science, LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)