Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

50231-U

Spherisorb® ODS1 (5 µm) HPLC Column

L × I.D. 12.5 cm × 4.6 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Spherisorb® ODS1 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4.6 mm

agency

suitable for USP L1

product line

Spherisorb®

feature

endcapped

manufacturer/tradename

Spherisorb®

extent of labeling

6.2% Carbon loading

parameter

20-45 °C temp. limit
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

12.5 cm × 4.6 mm

surface area

220 m2/g

matrix

octadecyl

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

80 Å

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Spherisorb is a registered trademark of Waters

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service