Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

502049

Supelco

EPA 8270/625/CLP/Appendix IX Semivolatile Calibration Mix

certified reference material, 1000 μg/mL each component in dichloromethane : benzene (3:1)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

certified reference material
TraceCERT®

agency

EPA 625,8270

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

1000 μg/mL each component in dichloromethane : benzene (3:1)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

-10 to -25°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Acenaphthene
    Acenaphthylene
    Anthracene
    Azobenzene
    Benz[a]anthracene
    Benzo[b]fluoranthene
    Benzo[k]fluoranthene
    Benzo[ghi]perylene
    Benzo[a]pyrene
    Benzyl butyl phthalate
    Bis(2-chloroethoxy)methane
    Bis(2-chloroethyl) ether
    Bis(2-ethylhexyl) phthalate
    4-Bromodiphenyl ether
    Carbazole
    4-Chlorodiphenyl ether
    Bis-(2-chloroisopropyl) ether
    2-Chloronaphthalene
    Chrysene
    Dibenz[a,h]anthracene
    Dibutyl phthalate
    1,2-Dichlorobenzene
    1,3-Dichlorobenzene
    1,4-Dichlorobenzene
    Diethyl phthalate
    Dimethyl phthalate
    2,4-Dinitrotoluene
    2,6-Dinitrotoluene
    Di-n-octyl phthalate
    Fluoranthene
    Fluorene
    Hexachlorobenzene
    Hexachloro-1,3-butadiene
    Hexachlorocyclopentadiene
    Hexachloroethane
    Indeno[1,2,3-cd]pyrene
    Isophorone
    Naphthalene
    Nitrobenzene
    N-Nitrosodimethylamine
    N-Nitrosodi-n-propylamine
    Phenanthrene
    Pyrene
    1,2,4-Trichlorobenzene
    See All (44)

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Blood, Central nervous system

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service